Assay Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177460.4 | NP_803411.3 | 2 - 3 | 459 | 97 | |
| GenBank mRNA | AK033791.1 | - | 409 | 97 | ||
| AK133475.1 | - | 2 - 3 | 460 | 97 | ||
| AK166002.1 | - | 1 - 2 | 223 | 97 | ||
| AK169866.1 | - | 2 - 3 | 374 | 97 | ||
More Information
Set Membership: |
|

Parp16
Genomic Map