Assay Details
| Gene Symbol: Zmym6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285885.1 | NP_001272814.1 | 6 - 7 | 1895 | 108 | |
| NM_001285888.1 | NP_001272817.1 | 7 - 8 | 1095 | 108 | ||
| NM_177462.5 | NP_803413.1 | 7 - 8 | 1088 | 108 | ||
| NR_104366.1 | 7 - 8 | 2151 | 108 | |||
| GenBank mRNA | AK035136.1 | - | 7 - 8 | 1090 | 108 | |
| AK082594.1 | - | 6 - 7 | 1619 | 108 | ||
| AK170695.1 | - | 7 - 8 | 2111 | 108 | ||
| BC034119.1 | - | 5 - 6 | 740 | 108 | ||
| BC040744.1 | - | 5 - 6 | 740 | 108 | ||
| BC094598.1 | - | 6 - 7 | 1868 | 108 | ||
| BC140958.1 | - | 7 - 8 | 963 | 9055570 | 108 | |
| BC144807.1 | - | 7 - 8 | 970 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zmym6
Genomic Map