Assay Details
| Gene Symbol: Arhgef15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177566.3 | NP_808234.2 | 4 - 5 | 1133 | 82 | |
| GenBank mRNA | AK051749.1 | - | 1 - 2 | 263 | 82 | |
| BC089365.1 | - | 4 - 5 | 1133 | 6844583 | 82 | |
| BC116343.1 | - | 4 - 5 | 1109 | 40098168 | 82 | |
| BC116344.1 | - | 4 - 5 | 1109 | 40098169 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051749.1 transcripts.
Set Membership: |
|

Arhgef15
Genomic Map