Assay Details
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177566.3 | NP_808234.2 | 5 - 6 | 1348 | 95 | |
GenBank mRNA | AK051749.1 | - | 2 - 3 | 478 | 95 | |
BC089365.1 | - | 5 - 6 | 1348 | 6844583 | 95 | |
BC116343.1 | - | 5 - 6 | 1324 | 40098168 | 95 | |
BC116344.1 | - | 5 - 6 | 1324 | 40098169 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK051749.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
