Assay Details
| Gene Symbol: D630003M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131021.2 | NP_001124493.1 | 4 - 5 | 1862 | 57 | |
| NM_177657.5 | NP_808325.2 | 4 - 5 | 1862 | 57 | ||
| GenBank mRNA | AK052605.1 | - | 4 - 5 | 1861 | 57 | |
| AK168367.1 | - | 4 - 5 | 1864 | 57 | ||
| BC137959.1 | - | 4 - 5 | 1773 | 8860978 | 57 | |
| BC137960.1 | - | 4 - 5 | 1773 | 8860979 | 57 | |
More Information
Set Membership: |
|

D630003M21Rik
Genomic Map