Assay Details
| Gene Symbol: Dnajc5g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177677.3 | NP_808345.1 | 1 - 2 | 98 | 70 | |
| GenBank mRNA | AK076638.1 | - | 1 - 2 | 99 | 70 | |
| BC089502.1 | - | 1 - 2 | 77 | 6774833 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076638.1, BC089502.1, NM_177677.3 transcripts.
Set Membership: |
|

Dnajc5g
Genomic Map