Assay Details
| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177688.4 | NP_808356.1 | 1 | 1524 | 63 | |
| GenBank mRNA | AK053755.1 | - | 1 | 1451 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2afj| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177688.4 | NP_808356.1 | 1 | 1524 | 63 | |
| GenBank mRNA | AK053755.1 | - | 1 | 1451 | 63 | |
Set Membership: |
|