Assay Details
| Gene Symbol: Nlrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177690.3 | NP_808358.2 | 10 - 11 | 2907 | 70 | |
| GenBank mRNA | AK054264.1 | - | 10 - 11 | 2908 | 70 | |
| AY360473.1 | - | 10 - 11 | 2908 | 70 | ||
| DQ066881.1 | - | 10 - 11 | 2908 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nlrp2
Genomic Map