Assay Details
| Gene Symbol: Nlrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177690.3 | NP_808358.2 | 4 - 5 | 459 | 106 | |
| GenBank mRNA | AK054264.1 | - | 4 - 5 | 460 | 106 | |
| AY360473.1 | - | 4 - 5 | 460 | 106 | ||
| DQ066881.1 | - | 4 - 5 | 460 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp2
Genomic Map