Assay Details
| Gene Symbol: Nlrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177690.3 | NP_808358.2 | 9 - 10 | 2740 | 120 | |
| GenBank mRNA | AK054264.1 | - | 9 - 10 | 2741 | 120 | |
| AY360473.1 | - | 9 - 10 | 2741 | 120 | ||
| DQ066881.1 | - | 9 - 10 | 2741 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042247 | AY360473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042247 | DQ066881.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042247 | NM_177690.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042247 | AK054264.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472949 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38373 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810651 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 7977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83821 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172824 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165049 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163086 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164473 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042247 | AY360473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042247 | DQ066881.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042247 | NM_177690.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042247 | AK054264.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24952750 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164473 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172824 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06115633 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06115587 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163086 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165049 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01476536 | - | - |
| Whole Mouse Genome G4122A | A_51_P181031 | AY360473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181031 | DQ066881.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181031 | NM_177690.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181031 | AK054264.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164473 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06115633 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163086 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172824 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06223914 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06223913 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06223908 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06223903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24952744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06485326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06223905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06223914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06446917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24952747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559731 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559730 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700020003.mm.1 | TC0700002191.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020003.mm.1 | NM_177690.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020003.mm.1 | NM_177690.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020002.mm.1 | TC0700002191.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020002.mm.1 | NM_177690.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020002.mm.1 | NM_177690.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 743362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3710451 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710451 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698602 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698602 | - | - |
| mouse-6 v1.1 (Illumina) | scl31783.12.1_220-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710451 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31783.12.1_220-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710451 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2698602 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698602 | - | - |

Nlrp2
Genomic Map