Assay Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073508.1 | 3 - 4 | 294 | 88 | ||
| GenBank mRNA | AK034197.1 | - | 3 - 4 | 283 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034197.1 transcripts.
Set Membership: |
|

Ano5
Genomic Map