Assay Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271879.1 | NP_001258808.1 | 6 - 7 | 777 | 71 | |
| NM_177694.6 | NP_808362.2 | 7 - 8 | 819 | 71 | ||
| NR_073508.1 | 7 - 8 | 812 | 71 | |||
| GenBank mRNA | AB125740.1 | - | 7 - 8 | 819 | 71 | |
| AB206762.1 | - | 7 - 8 | 819 | 71 | ||
| AK034197.1 | - | 7 - 8 | 801 | 71 | ||
| BC109163.2 | - | 6 - 7 | 699 | 40047987 | 71 | |
More Information
Set Membership: |
|

Ano5
Genomic Map