Assay Details
| Gene Symbol: Fhod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177699.4 | NP_808367.2 | 19 - 20 | 3265 | 81 | |
| GenBank mRNA | AB041045.1 | - | 19 - 20 | 3159 | 81 | |
| AK030737.1 | - | 19 - 20 | 3227 | 81 | ||
| BC060654.1 | - | 19 - 20 | 3248 | 30354865 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fhod1
Genomic Map