Assay Details
| Gene Symbol: Lsm14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177727.4 | NP_808395.2 | 6 - 7 | 1033 | 88 | |
| GenBank mRNA | AK035040.1 | - | 6 - 7 | 1062 | 88 | |
| AK162234.1 | - | 1 - 2 | 150 | 88 | ||
| BC040823.1 | - | 6 - 7 | 1033 | 88 | ||
| BC099395.1 | - | 3 - 4 | 452 | 30933544 | 88 | |
| BY720115.1 | - | 1 - 2 | 148 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsm14b
Genomic Map