Assay Details
| Gene Symbol: Lsm14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177727.4 | NP_808395.2 | 7 - 8 | 1184 | 102 | |
| GenBank mRNA | AK035040.1 | - | 7 - 8 | 1213 | 102 | |
| AK162234.1 | - | 2 - 3 | 301 | 102 | ||
| BC040823.1 | - | 7 - 8 | 1184 | 102 | ||
| BC099395.1 | - | 4 - 5 | 603 | 30933544 | 102 | |
| BY720115.1 | - | 2 - 3 | 299 | 102 | ||
| DV654874.1 | - | 1 - 2 | 133 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsm14b
Genomic Map