Assay Details
| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161665.1 | NP_001155137.1 | 13 - 14 | 6478 | 64 | |
| GenBank mRNA | AB355846.1 | - | 13 - 14 | 6478 | 64 | |
| AK076435.1 | - | 2 - 3 | 2223 | 64 | ||
| BC056349.1 | - | 8 - 9 | 4561 | 6405419 | 64 | |
| BC068141.1 | - | 10 - 11 | 5290 | 64 | ||
More Information
Set Membership: |
|

Kif26b
Genomic Map