Assay Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177765.3 | NP_808433.2 | 5 - 6 | 844 | 85 | |
| GenBank mRNA | AK029565.1 | - | 5 - 6 | 843 | 85 | |
| AK161519.1 | - | 5 - 6 | 667 | 85 | ||
| AM690757.1 | - | 4 - 5 | 545 | 85 | ||
| BC145749.1 | - | 4 - 5 | 452 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll13
Genomic Map