Assay Details
| Gene Symbol: Exoc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177788.4 | NP_808456.1 | 11 - 12 | 1985 | 89 | |
| GenBank mRNA | AK050100.1 | - | 11 - 12 | 1986 | 89 | |
| AK140526.1 | - | 7 - 8 | 1077 | 89 | ||
| AK172618.1 | - | 10 - 11 | 1652 | 89 | ||
| BC094238.1 | - | 11 - 12 | 1939 | 30604935 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc3l
Genomic Map