Assay Details
| Gene Symbol: Exoc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177788.4 | NP_808456.1 | 6 - 7 | 1399 | 102 | |
| GenBank mRNA | AK050100.1 | - | 6 - 7 | 1400 | 102 | |
| AK140526.1 | - | 2 - 3 | 491 | 102 | ||
| AK172618.1 | - | 5 - 6 | 1066 | 102 | ||
| BC094238.1 | - | 6 - 7 | 1353 | 30604935 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc3l
Genomic Map