Assay Details
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301173.1 | NP_001288102.1 | 13 - 14 | 2465 | 90 | |
NM_177791.3 | NP_808459.1 | 14 - 15 | 2534 | 90 | ||
GenBank mRNA | AK076644.1 | - | 14 - 15 | 2526 | 90 | |
BC092530.1 | - | 13 - 14 | 2465 | 6744085 | 90 | |
BC125431.1 | - | 14 - 15 | 2526 | 40129946 | 90 | |
BC125433.1 | - | 14 - 15 | 2526 | 40129948 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
