Assay Details
| Gene Symbol: Pxdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181395.2 | NP_852060.2 | 8 - 9 | 939 | 66 | |
| GenBank mRNA | AK054255.1 | - | 7 - 8 | 854 | 66 | |
| AK122223.1 | - | 8 - 9 | 709 | 66 | ||
| AK142872.1 | - | 8 - 9 | 939 | 66 | ||
| AK163597.1 | - | 8 - 9 | 926 | 66 | ||
| BC150789.1 | - | 8 - 9 | 934 | 9087700 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pxdn
Genomic Map