Assay Details
| Gene Symbol: Arhgef37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177828.4 | NP_808496.2 | 10 - 11 | 1624 | 76 | |
| GenBank mRNA | AB199795.1 | - | 9 - 10 | 1474 | 76 | |
| AK030242.1 | - | 6 - 7 | 1451 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef37
Genomic Map