Assay Details
| Gene Symbol: Cpa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289497.1 | NP_001276426.1 | 5 - 6 | 724 | 63 | |
| NM_177834.4 | NP_808502.2 | 6 - 7 | 849 | 63 | ||
| GenBank mRNA | AK078883.1 | - | 5 - 6 | 725 | 63 | |
| AY773477.1 | - | 6 - 7 | 847 | 63 | ||
| BC119559.1 | - | 6 - 7 | 834 | 40057538 | 63 | |
| BC119560.1 | - | 6 - 7 | 834 | 40057539 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpa6
Genomic Map