Assay Details
| Gene Symbol: Tnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177839.3 | NP_808507.2 | 13 - 14 | 3293 | 85 | |
| GenBank mRNA | AF455756.1 | - | 13 - 14 | 3260 | 85 | |
| AK048033.1 | - | 6 - 7 | 1408 | 85 | ||
| BC138335.1 | - | 13 - 14 | 3254 | 8861355 | 85 | |
| BC138336.1 | - | 13 - 14 | 3254 | 8861356 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnn
Genomic Map