Assay Details
| Gene Symbol: Dido1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291433.1 | NP_001278362.1 | 8 - 9 | 2533 | 91 | |
| NM_175551.4 | NP_780760.2 | 8 - 9 | 2533 | 91 | ||
| NM_177852.4 | NP_808520.2 | 8 - 9 | 2481 | 91 | ||
| GenBank mRNA | AK049707.1 | - | 2 - 3 | 1148 | 91 | |
| AK129117.1 | - | 8 - 9 | 2481 | 91 | ||
| AY425951.1 | - | 8 - 9 | 2435 | 91 | ||
| AY425952.1 | - | 8 - 9 | 2435 | 91 | ||
| BC044755.1 | - | 2 - 3 | 632 | 91 | ||
| BC138712.1 | - | 7 - 8 | 2310 | 8861734 | 91 | |
| BC138713.1 | - | 7 - 8 | 2310 | 8861735 | 91 | |
More Information
Set Membership: |
|

Dido1
Genomic Map