Assay Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177859.3 | NP_808527.2 | 3 - 4 | 1331 | 82 | |
| GenBank mRNA | AK029552.1 | - | 3 - 4 | 1209 | 82 | |
| AK135678.1 | - | 2 - 3 | 291 | 82 | ||
| AK161171.1 | - | 2 - 3 | 163 | 82 | ||
| AK164420.1 | - | 3 - 4 | 1334 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164420.1 transcripts.
Set Membership: |
|

Aknad1
Genomic Map