Assay Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177859.3 | NP_808527.2 | 8 - 9 | 1893 | 134 | |
| GenBank mRNA | AK029552.1 | - | 9 - 10 | 2063 | 134 | |
| AK135678.1 | - | 7 - 8 | 853 | 134 | ||
| AK164420.1 | - | 8 - 9 | 1896 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155776 | AK029552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155776 | AK135678.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155776 | AK161171.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155776 | AK164420.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155776 | NM_177859.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2179691 | NM_177859.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | AK146840.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | AK166187.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | AK167423.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | AK168165.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | AK171238.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | BC011110.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | BC071220.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | BC080791.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | BC092072.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | CT010192.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | D12780.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | NM_007444.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | NM_009665.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | XM_001476532.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | XM_001479341.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | XM_001480145.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894994 | Z14986.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892317 | XM_620643.4 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807376 | AK040451.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28126 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166746 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172748 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144922 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155776 | AK029552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155776 | AK135678.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155776 | AK161171.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155776 | AK164420.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155776 | NM_177859.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2179691 | NM_177859.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00723924 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166746 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172748 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05026041 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144922 | - | - |
| Whole Mouse Genome G4122A | A_52_P527899 | AK029552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527899 | AK135678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527899 | AK164420.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527899 | NM_177859.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P358127 | AK135678.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P358127 | AK164420.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P358127 | NM_177859.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129014 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126197 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144922 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166746 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02387511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02363438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02574322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03082658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02981512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02981514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02373151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02976260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02574317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02820318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02632747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03130378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02387498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02560824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03146422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02363441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02598366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03074121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03049608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02812551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02513024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02560833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02663451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02756032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02574328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02665787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02405933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02373152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02561903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22414501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02598357 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495377 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495381 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495380 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495378 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495374 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495372 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495379 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440249_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495371 | - | - |
| Mouse Genome 430 2.0 Array | 1440249_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440249_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329738_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329738_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300009448.mm.1 | NM_177859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009448.mm.1 | NM_177859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009448.mm.1 | TC0300001154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009444.mm.1 | NM_177859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009444.mm.1 | NM_177859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009444.mm.1 | TC0300001154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009445.mm.1 | NM_177859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009445.mm.1 | NM_177859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009445.mm.1 | TC0300001154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009443.mm.1 | NM_177859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009443.mm.1 | NM_177859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009443.mm.1 | TC0300001154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009447.mm.1 | NM_177859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009447.mm.1 | NM_177859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009447.mm.1 | TC0300001154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300009449.mm.1 | NM_177859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009449.mm.1 | NM_177859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300009449.mm.1 | TC0300001154.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440249_at | - | - |
| Mouse Expression Set 430 | 1440249_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440249_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440249_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 319400 | - | - |
| ABI Mouse Genome Survey Microarray | 411043 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 70551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2627377 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2627377 | - | - |
| mouse-6 v1.1 (Illumina) | scl22715.9.1_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22715.9.1_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2627377 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2627377 | - | - |

Aknad1
Genomic Map