Assay Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177866.4 | NP_808534.1 | 4 - 5 | 434 | 121 | |
| GenBank mRNA | AK077145.1 | - | 4 - 5 | 421 | 121 | |
| BC108974.2 | - | 4 - 5 | 414 | 40047424 | 121 | |
| BC108975.1 | - | 4 - 5 | 414 | 40047425 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
sodium ion transport
calcium ion transport
multicellular organism development
spermatogenesis
cell differentiation
sperm motility
regulation of ion transmembrane transport
sperm capacitation
transmembrane transport
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
ion channel activity
calcium activated cation channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel activity
protein binding
transport
ion transport
sodium ion transport
calcium ion transport
multicellular organism development
spermatogenesis
cell differentiation
sperm motility
regulation of ion transmembrane transport
sperm capacitation
transmembrane transport
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
ion channel activity
calcium activated cation channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel activity
protein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994284 | NM_001130030.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994284 | NM_177866.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994284 | AK077145.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050897 | BC051630.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130718 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104560 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177959 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994284 | NM_001130030.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994284 | NM_177866.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994284 | AK077145.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234440 | BC108974.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234440 | BC108975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234440 | EU781510.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234440 | EU781511.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234440 | NM_001130030.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23321655 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00994374 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177959 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104560 | - | - |
| Whole Mouse Genome G4122A | A_51_P234440 | BC108974.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234440 | BC108975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234440 | EU781510.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234440 | EU781511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234440 | NM_001130030.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108952 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109906 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177959 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104560 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03884470 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03884464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03884439 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03884460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03884455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23321703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03884460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03467002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03566601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03389418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03341665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03341666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04098679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03510856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03993688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03923106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23321662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03626732 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517212 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517206 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517199 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400030668.mm.1 | TC0400003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030668.mm.1 | NM_177866.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030668.mm.1 | NM_177866.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030669.mm.1 | TC0400003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030669.mm.1 | NM_177866.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030669.mm.1 | NM_177866.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030670.mm.1 | TC0400003730.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030670.mm.1 | NM_001130030.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030670.mm.1 | NM_001130030.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030670.mm.1 | NM_177866.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030670.mm.1 | NM_177866.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3440181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440181 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2591842 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2591842 | - | - |
| mouse-6 v1.1 (Illumina) | scl23695.12.1_184-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440181 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23695.12.1_184-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440181 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2991970 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2991970 | - | - |

Catsper4
Genomic Map