Assay Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177866.4 | NP_808534.1 | 6 - 7 | 649 | 96 | |
| GenBank mRNA | AK077145.1 | - | 6 - 7 | 636 | 96 | |
| BC108974.2 | - | 6 - 7 | 629 | 40047424 | 96 | |
| BC108975.1 | - | 6 - 7 | 629 | 40047425 | 96 | |
| EU781510.1 | - | 5 - 6 | 526 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Catsper4
Genomic Map