Assay Details
| Gene Symbol: 4933411G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177880.4 | NP_808548.1 | 1 - 2 | 534 | 78 | |
| GenBank mRNA | AK030175.1 | - | 1 - 2 | 535 | 78 | |
| BC115568.1 | - | 1 - 2 | 67 | 40102090 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030175.1, BC115568.1, NM_177880.4 transcripts.
Set Membership: |
|

4933411G11Rik
Genomic Map