| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600017399.mm.1 |
TC0600002214.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017399.mm.1 |
NM_177882.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017399.mm.1 |
NM_177882.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017397.mm.1 |
TC0600002214.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017397.mm.1 |
NM_177882.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017397.mm.1 |
NM_177882.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017398.mm.1 |
TC0600002214.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017398.mm.1 |
NM_177882.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017398.mm.1 |
NM_177882.4 |
Refseq |