Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142781.1 | NP_001136253.1 | 1 - 2 | 185 | 102 | |
| NM_177883.4 | NP_808551.2 | 1 - 2 | 185 | 102 | ||
| GenBank mRNA | AK082855.1 | - | 1 - 2 | 186 | 102 | |
| AK142385.1 | - | 1 - 2 | 103 | 102 | ||
| BC118939.1 | - | 1 - 2 | 179 | 102 | ||
| BC118940.1 | - | 1 - 2 | 179 | 40092089 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082855.1, AK142385.1, BC118940.1, NM_001142781.1, NM_177883.4 transcripts.
Set Membership: |
|

Fam188b
Genomic Map