Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142781.1 | NP_001136253.1 | 2 - 3 | 309 | 78 | |
| NM_177883.4 | NP_808551.2 | 2 - 3 | 309 | 78 | ||
| GenBank mRNA | AK082855.1 | - | 2 - 3 | 310 | 78 | |
| AK142385.1 | - | 2 - 3 | 227 | 78 | ||
| AK161304.1 | - | 2 - 3 | 189 | 78 | ||
| BC118939.1 | - | 2 - 3 | 303 | 78 | ||
| BC118940.1 | - | 2 - 3 | 303 | 40092089 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam188b
Genomic Map