| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600005096.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005096.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005096.mm.1 |
NM_001142781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005096.mm.1 |
NM_177883.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005096.mm.1 |
NM_177883.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005097.mm.1 |
TC0600000663.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005097.mm.1 |
NM_001142781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005097.mm.1 |
NM_001142781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005097.mm.1 |
NM_177883.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005097.mm.1 |
NM_177883.4 |
Refseq |