Assay Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177884.3 | NP_808552.2 | 10 - 11 | 1538 | 82 | |
| GenBank mRNA | AK038369.1 | - | 11 - 12 | 1525 | 82 | |
| AK050789.1 | - | 10 - 11 | 1480 | 82 | ||
| AK078171.1 | - | 10 - 11 | 1529 | 82 | ||
| AK136387.1 | - | 8 - 9 | 1140 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcfc2
Genomic Map