Assay Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177884.3 | NP_808552.2 | 1 - 2 | 272 | 102 | |
| GenBank mRNA | AK038369.1 | - | 1 - 2 | 269 | 102 | |
| AK050789.1 | - | 1 - 2 | 214 | 102 | ||
| AK077263.1 | - | 1 - 2 | 262 | 102 | ||
| AK078171.1 | - | 1 - 2 | 263 | 102 | ||
| BY730207.1 | - | 1 - 2 | 272 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050789.1 transcripts.
Set Membership: |
|

Gcfc2
Genomic Map