| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
TC0700001364.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045969.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045969.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045971.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045971.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045972.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011605.mm.1 |
NR_045972.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
TC0700001364.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045969.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045969.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045971.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045971.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045972.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700011604.mm.1 |
NR_045972.1 |
Refseq |