Assay Details
| Gene Symbol: B4galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177897.3 | NP_808565.2 | 16 - 17 | 2634 | 58 | |
| GenBank mRNA | AB114827.1 | - | 16 - 17 | 2634 | 58 | |
| BC038881.1 | - | 2 - 3 | 581 | 58 | ||
| BC062653.1 | - | 12 - 13 | 2324 | 58 | ||
| BC079533.1 | - | 5 - 6 | 1537 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galnt4
Genomic Map