Assay Details
Gene Symbol: Zfp866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177899.3 | NP_808567.2 | 3 - 4 | 306 | 118 | |
GenBank mRNA | AK052363.1 | - | 3 - 4 | 285 | 118 | |
AK133399.1 | - | 371 | 118 | |||
AK137334.1 | - | 3 - 4 | 308 | 118 | ||
AK140357.1 | - | 3 - 4 | 301 | 118 | ||
BC057095.1 | - | 3 - 4 | 280 | 6419770 | 118 | |
BC117846.1 | - | 3 - 4 | 261 | 40093653 | 118 | |
BC117847.1 | - | 3 - 4 | 261 | 40093655 | 118 | |
BC138224.1 | - | 3 - 4 | 257 | 8861243 | 118 | |
BC138227.1 | - | 3 - 4 | 257 | 8861246 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947916 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947917 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947915 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947914 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P22781 | CO042014.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P22781 | AK133399.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P22781 | NM_177899.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P22781 | CO042014.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P22781 | AK133399.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P22781 | NM_177899.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC138227.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC117846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC117847.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC138224.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | AK140357.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC138227.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC117846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC117847.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | BC138224.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P493322 | AK140357.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022885 | AK140357.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022885 | BC057095.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022885 | AK137334.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022885 | NM_177899.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022885 | AK052363.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722497 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701677 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262062 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162739 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850271 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941015 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402404 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793905 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608632 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479704 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374433 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772737 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608339 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422344 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9471 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164964 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49872 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161150 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3891 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97348 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105435 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44915 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163914 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96177 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126910 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156271 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153143 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P22781 | CO042014.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P22781 | AK133399.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P22781 | NM_177899.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P22781 | CO042014.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P22781 | AK133399.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P22781 | NM_177899.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC138227.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC117846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC117847.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC138224.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | AK140357.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC138227.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC117846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC117847.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | BC138224.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P493322 | AK140357.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022885 | AK140357.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022885 | BC057095.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022885 | AK137334.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022885 | NM_177899.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2022885 | AK052363.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947920 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947909 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947917 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947926 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25947912 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153143 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01761898 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126910 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01761881 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06546656 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156271 | - | - |
Whole Mouse Genome G4122A | A_52_P22781 | CO042014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P22781 | AK133399.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P22781 | NM_177899.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P22781 | CO042014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P22781 | AK133399.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P22781 | NM_177899.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC138227.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC117846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC117847.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC138224.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | AK140357.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC138227.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC117846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC117847.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | BC138224.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P493322 | AK140357.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129929 | BC138227.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129929 | BC117846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129929 | BC117847.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129929 | BC138224.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P129929 | AK140357.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139253 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153143 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126910 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156271 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07123854 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07311725 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07311730 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07311727 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947914 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947917 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947915 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25947916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25947925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06873615 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07157491 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07311724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947923 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07110496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947892 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06739989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07311727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07117622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25947845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06836123 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10579093 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579095 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579094 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579092 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579100 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579098 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579099 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579097 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10579096 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455334_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10579089 | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330788_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330788_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800017853.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017853.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017853.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017857.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017857.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017857.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017866.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017866.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017866.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017855.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017855.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017855.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017861.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017861.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017861.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017852.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017852.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017852.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017854.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017854.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017854.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017858.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017858.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017858.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017859.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017859.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017859.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017862.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017862.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017862.mm.1 | TC0800002372.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800017860.mm.1 | NM_177899.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017860.mm.1 | NM_177899.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800017860.mm.1 | TC0800002372.mm.1 | MTA TC |
Murine Genome U74v2 Set | 170206_PM_at | - | - |
Murine Genome U74v2 Set | 170206_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455334_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455334_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2640707 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002640707 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769672 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2769672 | - | - |
mouse-6 v1.1 (Illumina) | scl0330788.1_4-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2640707 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0330788.1_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640707 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769672 | - | - |
