Assay Details
| Gene Symbol: Slc9a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177909.5 | NP_808577.3 | 8 - 9 | 1166 | 90 | |
| GenBank mRNA | AK037058.1 | - | 8 - 9 | 1150 | 90 | |
| AK156494.1 | - | 8 - 9 | 1127 | 90 | ||
| AK172687.1 | - | 8 - 9 | 1138 | 90 | ||
| BC099942.1 | - | 8 - 9 | 1140 | 30103308 | 90 | |
| BC153825.1 | - | 8 - 9 | 1144 | 40093280 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a9
Genomic Map