Assay Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178065.4 | NP_835166.3 | 12 - 13 | 2133 | 101 | |
| GenBank mRNA | AB093230.1 | - | 11 - 12 | 1752 | 101 | |
| AK077015.1 | - | 11 - 12 | 1665 | 101 | ||
| AK133332.1 | - | 11 - 12 | 1669 | 101 | ||
| AK147491.1 | - | 12 - 13 | 2134 | 101 | ||
| AK147878.1 | - | 12 - 13 | 2097 | 101 | ||
| AK154529.1 | - | 12 - 13 | 2114 | 101 | ||
| BC006074.1 | - | 3 - 4 | 288 | 101 | ||
| BC049900.1 | - | 6 - 7 | 823 | 101 | ||
| BC060658.1 | - | 11 - 12 | 1702 | 30355406 | 101 | |
| BC079590.1 | - | 12 - 13 | 2091 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arel1
Genomic Map