Assay Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178065.4 | NP_835166.3 | 16 - 17 | 2675 | 79 | |
| GenBank mRNA | AB093230.1 | - | 15 - 16 | 2294 | 79 | |
| AK077015.1 | - | 15 - 16 | 2207 | 79 | ||
| AK133332.1 | - | 15 - 16 | 2211 | 79 | ||
| AK147491.1 | - | 16 - 17 | 2676 | 79 | ||
| AK147878.1 | - | 16 - 17 | 2639 | 79 | ||
| AK154529.1 | - | 16 - 17 | 2656 | 79 | ||
| BC006074.1 | - | 7 - 8 | 830 | 79 | ||
| BC049900.1 | - | 10 - 11 | 1365 | 79 | ||
| BC057351.1 | - | 15 - 16 | 2503 | 79 | ||
| BC060658.1 | - | 15 - 16 | 2244 | 30355406 | 79 | |
| BC079590.1 | - | 16 - 17 | 2629 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arel1
Genomic Map