Assay Details
| Gene Symbol: Rbm45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153405.2 | NP_700454.1 | 2 - 3 | 566 | 158 | |
| GenBank mRNA | AB036992.1 | - | 2 - 3 | 482 | 158 | |
| AK080420.1 | - | 2 - 3 | 540 | 158 | ||
| AK089975.1 | - | 2 - 3 | 459 | 158 | ||
| AK090084.1 | - | 2 - 3 | 535 | 158 | ||
| AK140861.1 | - | 2 - 3 | 567 | 158 | ||
| AY152391.1 | - | 2 - 3 | 566 | 158 | ||
| BC057890.1 | - | 2 - 3 | 477 | 4979982 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm45
Genomic Map