Assay Details
| Gene Symbol: Camta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190376.1 | NP_001177305.1 | 7 - 8 | 845 | 67 | |
| NM_001190378.1 | NP_001177307.1 | 7 - 8 | 845 | 67 | ||
| NM_001190379.1 | NP_001177308.1 | 6 - 7 | 773 | 67 | ||
| NM_178116.4 | NP_835217.1 | 7 - 8 | 860 | 67 | ||
| GenBank mRNA | AK039670.1 | - | 7 - 8 | 672 | 67 | |
| AK122400.1 | - | 5 - 6 | 501 | 67 | ||
| AK150186.1 | - | 7 - 8 | 697 | 67 | ||
| AK169118.1 | - | 6 - 7 | 612 | 67 | ||
| BC023962.1 | - | 2 - 3 | 659 | 67 | ||
| BC049133.1 | - | 7 - 8 | 689 | 6827800 | 67 | |
| BC056395.1 | - | 7 - 8 | 674 | 6852349 | 67 | |
| BC059065.1 | - | 6 - 7 | 640 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Camta2
Genomic Map