Assay Details
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178240.2 | NP_839971.1 | 1 - 2 | 403 | 70 | |
GenBank mRNA | AY081947.1 | - | 1 - 2 | 403 | 70 | |
BC130022.1 | - | 1 - 2 | 406 | 40140742 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
