Assay Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 13 - 14 | 4847 | 73 | |
| NM_178242.2 | NP_839973.1 | 14 - 15 | 4976 | 73 | ||
| GenBank mRNA | AK141849.1 | - | 10 - 11 | 3945 | 73 | |
| AK173280.1 | - | 10 - 11 | 2605 | 73 | ||
| BC049818.1 | - | 14 - 15 | 4976 | 6511384 | 73 | |
More Information
Set Membership: |
|

Tnrc18
Genomic Map