Assay Details
| Gene Symbol: Teddm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178243.3 | NP_839974.2 | 1 | 189 | 66 | |
| GenBank mRNA | AJ554213.1 | - | 1 | 146 | 66 | |
| AK136655.1 | - | 1 | 191 | 66 | ||
| AK136685.1 | - | 1 | 190 | 66 | ||
| BC120491.1 | - | 1 | 143 | 40129414 | 66 | |
| DQ402363.1 | - | 1 | 189 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | DQ402363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | BC120491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | BC131905.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | AJ554213.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | AK136655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | DQ402363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | BC120491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | BC131905.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | AJ554213.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P85329 | AK136655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148519 | CR516116.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148519 | NM_178243.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492655 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158494 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128140 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | DQ402363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | BC120491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | BC131905.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | AJ554213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | AK136655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | DQ402363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | BC120491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | BC131905.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | AJ554213.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P85329 | AK136655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148519 | CR516116.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148519 | NM_178243.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20728986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20729012 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128140 | - | - |
| Whole Mouse Genome G4122A | A_52_P85329 | DQ402363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | BC120491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | BC131905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | AJ554213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | AK136655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | DQ402363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | BC120491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | BC131905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | AJ554213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P85329 | AK136655.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136011 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00674728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00674727 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00674719 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00173308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20729002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20728986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00939686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00674730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00566669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00197167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00358702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00477842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00349002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00401552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00896950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00348992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00674732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00173309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01089459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00338950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20728962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20729012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10358923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10358925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10358924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10358922 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444658_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10358921 | - | - |
| Mouse Genome 430 2.0 Array | 1444658_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444658_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240817_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240817_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100026747.mm.1 | NM_178243.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026747.mm.1 | NM_178243.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026747.mm.1 | TC0100003283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026745.mm.1 | NM_178243.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026745.mm.1 | NM_178243.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026745.mm.1 | TC0100003283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100026744.mm.1 | NM_178243.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026744.mm.1 | NM_178243.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100026744.mm.1 | TC0100003283.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1444658_PM_at | - | - |
| Mouse Expression Set 430 | 1444658_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444658_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444658_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3170398 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237653 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237653 | - | - |
| mouse-6 v1.1 (Illumina) | scl0240817.1_172-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0240817.1_172-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237653 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237653 | - | - |

Teddm2
Genomic Map