Assay Details
| Gene Symbol: Nrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK051824.1 | - | 1 - 2 | 484 | 109 | |
| AY648976.1 | - | 1 - 2 | 101 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Nrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK051824.1 | - | 1 - 2 | 484 | 109 | |
| AY648976.1 | - | 1 - 2 | 101 | 109 | ||
Set Membership: |
|