Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177324.2 | NP_796298.2 | 17 - 18 | 2078 | 90 | |
| GenBank mRNA | AK046622.1 | - | 17 - 18 | 2066 | 90 | |
| AK048952.1 | - | 17 - 18 | 2055 | 90 | ||
| AK080952.1 | - | 17 - 18 | 2079 | 90 | ||
| AK080963.1 | - | 17 - 18 | 2073 | 90 | ||
| AK087396.1 | - | 17 - 18 | 2073 | 90 | ||
| BC067204.1 | - | 18 - 19 | 2116 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sbf2
Genomic Map