Assay Details
| Gene Symbol: Prkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252458.1 | NP_001239387.1 | 14 - 15 | 2085 | 70 | |
| NM_178900.4 | NP_849231.1 | 13 - 14 | 2370 | 70 | ||
| GenBank mRNA | AK037030.1 | - | 13 - 14 | 2371 | 70 | |
| AK154029.1 | - | 13 - 14 | 2219 | 70 | ||
| AK170761.1 | - | 14 - 15 | 2049 | 70 | ||
| BC095949.1 | - | 14 - 15 | 2035 | 4950957 | 70 | |
| BC096444.1 | - | 14 - 15 | 2053 | 4216827 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd2
Genomic Map